Learn More
Invitrogen™ CHD7 Polyclonal Antibody

Description
Immunogen sequence: ALNLSVPRQRR RRRRKIEIEA ERAAKRRNLM EMVAQLRESQ VVSENGQEKV VDLSKASREA TSSTSNFSSL SSKFILPNVS TPVSDAFKTQ MEL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 98%.
Specifications
Specifications
| Antigen | CHD7 |
| Applications | ChIP Assay, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CHD7 |
| Gene Accession No. | Q9P2D1 |
| Gene Alias | A730019I05Rik; ATP-dependent helicase CHD7; CHARGE association; CHD7; CHD-7; chromodomain helicase DNA binding protein 7; chromodomain helicase DNA binding protein 7 isoform CRA_e; chromodomain-helicase-DNA-binding protein 7; CRG; Cycn; Cyn; Dz; Edy; Flo; GENA 47; Gena 52; GENA 60; HH5; IS3; KAL5; KIAA1416; Lda; metis; Mt; Obt; Todo; WBE1; Whi |
| Gene Symbols | CHD7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.