Learn More
CHD7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301222A
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that contains several helicase family domains. Mutations in this gene have been found in some patients with the CHARGE syndrome.Specifications
| CHD7 | |
| Polyclonal | |
| Unconjugated | |
| CHD7 | |
| HH5, IS3, KAL5, ATP-dependent helicase CHD7; chromodomain helicase DNA binding protein 7 isoform CRA_e; chromodomain-helicase-DNA-binding protein 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55636 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9P2D1 | |
| CHD7 | |
| Between 2025 and 2075 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.