Learn More
CHD7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier:  Bethyl Laboratories, Inc A301223A
Encompass_Preferred
Catalog No. 50-155-9818
Only null left
Add to Cart
Description
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that contains several helicase family domains. Mutations in this gene have been found in some patients with the CHARGE syndrome.
Specifications
Specifications
CHD7
Polyclonal
Unconjugated
CHD7
HH5, IS3, KAL5, ATP-dependent helicase CHD7; chromodomain helicase DNA binding protein 7 isoform CRA_e; chromodomain-helicase-DNA-binding protein 7
Rabbit
Antigen affinity chromatography
RUO
55636
4° C
Liquid
ChIP sequencing (ChIP-seq), Immunoprecipitation, Western Blot
0.2 mg/ml
TBS with 0.1% BSA and 0.09% sodium azide
Q9P2D1
CHD7
Between 2947 and 2997
100 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.