Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565706 Supplier Invitrogen™ Supplier No. PA565706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEFAVDPRFLA YMEDRRKQKW QRCKKNNKAE LNCLGMEPVQ TANSRNGKKG HHTETVFNRV LPGPIAPESS KKRARRMRPD LSKMMALMQG GSTGSLSLHN TFQHSSSGLQ SVSSLGHSSA TSASLPFMPF VMGGAPSSPH VD Highest antigen sequence identity to the following orthologs - mouse 90%, rat 91%.

DNA helicase that acts as a chromatin remodeling factor and regulates transcription. Acts as a transcription repressor by remodeling chromatin structure and recruiting histone H1 to target genes. Suppresses p53/TP53-mediated apoptosis by recruiting histone H1 and preventing p53/TP53 transactivation activity. Acts as a negative regulator of Wnt signaling pathway by regulating beta-catenin (CTNNB1) activity. Negatively regulates CTNNB1-targeted gene expression by being recruited specifically to the promoter regions of several CTNNB1 responsive genes. Involved in both enhancer blocking and epigenetic remodeling at chromatin boundary via its interaction with CTCF. Acts as a suppressor of STAT3 activity by suppressing the LIF-induced STAT3 transcriptional activity. Also acts as a transcription activator via its interaction with ZNF143 by participating in efficient U6 RNA polymerase III transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD8
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Chd8
Gene Accession No. Q9HCK8
Gene Alias 5830451P18Rik; ATP-dependent helicase CHD8; AU015341; AUTS18; Axis duplication inhibitor; beta-catenin binding protein; CHD8; CHD-8; chromodomain helicase DNA binding protein 8; chromodomain-helicase-DNA-binding protein 8; Duplin; helicase with SNF2 domain 1; HELSNF1; Kiaa1564; mKIAA1564
Gene Symbols Chd8
Host Species Rabbit
Immunogen Recombinant Human CHD8. Recombinant protein control fragment (Product #RP-106379).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57680
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.