Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD8 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | CHD8 |
---|---|
Dilution | Western Blot 0.04 to 0.4 μg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CHD8 Polyclonal antibody specifically detects CHD8 in Human samples. It is validated for Western BlotSpecifications
CHD8 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Wnt Signaling Pathway | |
PBS, pH 7.2, 40% glycerol | |
57680 | |
IgG | |
Affinity purified |
Western Blot 0.04 to 0.4 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ATP-dependent helicase CHD8, CHD-8, chromodomain helicase DNA binding protein 8, DKFZp686N17164, duplin, EC 3.6.1, EC 3.6.1.7, EC 3.6.4.12, Helicase with SNF2 domain 1chromodomain-helicase-DNA-binding protein 8, HELSNF1, KIAA1564axis duplication inhibitor | |
This antibody was developed against Recombinant Protein corresponding to amino acids: LDSLTDDSFNQVTQDPIEEALGLPSSLDSLDQMNQDGGGGDVGNSSASELVPPPEEAAPTELSKESTAPAPESITLHDYTTQPASQEQP | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title