Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD9 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060419AF594
Description
CHD9 Polyclonal antibody specifically detects CHD9 in Human samples. It is validated for Western Blot, ImmunoblotSpecifications
CHD9 | |
Polyclonal | |
Western Blot, Immunoblotting | |
AD013, ATP-dependent helicase CHD9, BC022889, CHD-9, chromatin remodeling factor CHROM1, Chromatin-related mesenchymal modulator, Chromatin-remodeling factor CHROM1, chromodomain helicase DNA binding protein 9, chromodomain-helicase-DNA-binding protein 9, ciprofibrate bound protein p240, CReMM, EC 3.6.1, EC 3.6.4.12, FLJ12178, KIAA0308, KISH2, Kismet homolog 2, Peroxisomal proliferator-activated receptor A-interacting complex 320 kDaprotein, PPAR{gamma}-interacting cofactor 320 kDa, PPAR-alpha-interacting complex protein 320 kDa, PRIC320, proteinx0008 | |
The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human chromodomain helicase DNA binding protein 9 (chromatin-related mesenchymal modulator) using the numbering given in entry AAQ24287.1 (GeneID 80205). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoblot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
80205 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction