Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHD9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103742 Supplier Invitrogen™ Supplier No. PA5103742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CHD9.

CHD9 (chromodomain-helicase-DNA-binding protein 9), also known as chromatin-related mesenchymal modulator (CReMM), PPAR-α-interacting complex protein, kismet homolog 2 or CHROM1, is a 2,897 amino acid protein belonging to the Snf2/Rad54 helicase family. The CHD family of proteins are ATP-dependent chromatin remodeling enzymes which combine chromodomains with SWI2/Snf2 ATPase/helicase motifs and DNA-binding capability. Localized to the cytoplasm and the nucleus, CHD9 contains two chromodomains, one ATPbinding helicase domain and one C-terminal helicase domain. Chromodomains are protein regions of about 40-50 amino acid residues found in proteins associated with chromatin remodeling and manipulation. The domain is highly conserved among both plants and animals and is found in a large variety of proteins from many genomes. CHD9 acts as a transcriptional coactivator for PPARα and may also be an ATP-dependent chromatin remodeling protein. CHD9 is widely expressed at low levels and is present as three isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Chd9
Gene Accession No. Q3L8U1, Q8BYH8
Gene Alias 1810014J18Rik; 9030205D12Rik; A330063D19Rik; AD013; AD-013; ATP-dependent helicase CHD9; BC022889; Chd9; CHD-9; chromatin remodeling factor CHROM1; chromatin-related mesenchymal modulator; Chromatin-remodeling factor CHROM1; chromodomain helicase DNA binding protein 9; chromodomain-helicase-DNA-binding protein 9; ciprofibrate bound protein p240; ciprofibrate-bound protein PRIC320; CReMM; FLJ12178; Kiaa0308; KISH2; Kismet homolog 2; mKIAA0308; peroxisomal proliferator-activated receptor A-interacting complex 320 kDa protein; PPAR{gamma}-interacting cofactor 320 kDa; PPAR-alpha-interacting complex protein 320 kDa; PRIC320; proteinx0008; x0008
Gene Symbols Chd9
Host Species Rabbit
Immunogen A synthesized peptide derived from human CHD9(Accession Q3L8U1), corresponding to amino acid residues I1450-K1500.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109151, 80205
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.