Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHD9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139877 Supplier Invitrogen™ Supplier No. PA5139877
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SLRSQQNRNN LNPGQNSLSQ SKNFMNVSGP HRVNVNHPPQ MTNASNSQQS ISMQQFSQTS NPSAHFHKCS SHQEGNFNGP SPNMTSCSVS NSQQFSSHYS FSSNHISPNS LLQSSA Highest antigen sequence indentity to the following orthologs: Mouse - 72%, Rat - 69%.

CHD9 (chromodomain-helicase-DNA-binding protein 9), also known as chromatin-related mesenchymal modulator (CReMM), PPAR-α-interacting complex protein, kismet homolog 2 or CHROM1, is a 2,897 amino acid protein belonging to the Snf2/Rad54 helicase family. The CHD family of proteins are ATP-dependent chromatin remodeling enzymes which combine chromodomains with SWI2/Snf2 ATPase/helicase motifs and DNA-binding capability. Localized to the cytoplasm and the nucleus, CHD9 contains two chromodomains, one ATPbinding helicase domain and one C-terminal helicase domain. Chromodomains are protein regions of about 40-50 amino acid residues found in proteins associated with chromatin remodeling and manipulation. The domain is highly conserved among both plants and animals and is found in a large variety of proteins from many genomes. CHD9 acts as a transcriptional coactivator for PPARα and may also be an ATP-dependent chromatin remodeling protein. CHD9 is widely expressed at low levels and is present as three isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD9
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Chd9
Gene Accession No. Q3L8U1
Gene Alias 1810014J18Rik; 9030205D12Rik; A330063D19Rik; AD013; AD-013; ATP-dependent helicase CHD9; BC022889; Chd9; CHD-9; chromatin remodeling factor CHROM1; chromatin-related mesenchymal modulator; Chromatin-remodeling factor CHROM1; chromodomain helicase DNA binding protein 9; chromodomain-helicase-DNA-binding protein 9; ciprofibrate bound protein p240; ciprofibrate-bound protein PRIC320; CReMM; FLJ12178; Kiaa0308; KISH2; Kismet homolog 2; mKIAA0308; peroxisomal proliferator-activated receptor A-interacting complex 320 kDa protein; PPAR{gamma}-interacting cofactor 320 kDa; PPAR-alpha-interacting complex protein 320 kDa; PRIC320; proteinx0008; x0008
Gene Symbols Chd9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHD9. Recombinant protein control fragment (Product #RP-109100).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80205
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.