Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD9 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | CHD9 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CHD9 Polyclonal antibody specifically detects CHD9 in Human samples. It is validated for ImmunofluorescenceSpecifications
CHD9 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
AD013, ATP-dependent helicase CHD9, BC022889, CHD-9, chromatin remodeling factor CHROM1, Chromatin-related mesenchymal modulator, Chromatin-remodeling factor CHROM1, chromodomain helicase DNA binding protein 9, chromodomain-helicase-DNA-binding protein 9, ciprofibrate bound protein p240, CReMM, EC 3.6.1, EC 3.6.4.12, FLJ12178, KIAA0308, KISH2, Kismet homolog 2, Peroxisomal proliferator-activated receptor A-interacting complex 320 kDaprotein, PPAR{gamma}-interacting cofactor 320 kDa, PPAR-alpha-interacting complex protein 320 kDa, PRIC320, proteinx0008 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: SLRSQQNRNNLNPGQNSLSQSKNFMNVSGPHRVNVNHPPQMTNASNSQQSISMQQFSQTSNPSAHFHKCSSHQEGNFNGPSPNMTSCSVSNSQQFSSHYSFSSNHISPNSLLQSSA | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
80205 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title