Learn More
Invitrogen™ CHDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595792
Description
Immunogen sequence: LHPALSRTNL KAEAETLVSR VLFEGTRAVG VEYVKNGQSH RAYASKEVIL SGGAINSPQL LMLSGIGNAD DLKKLGIPVV CHLPGVGQNL QDHLEIYIQQ ACTRPITLHS AQKPLRKVCI GLEWLWKFTG EGATAHLETG GFIRSQPGVP HPDIQFHFLP SQVIDHGRVP TQQEAYQVHV GPMRGTSVGW LKLRSANPQD HPVIQPNYLS TETDIEDFRL CVKLTREIFA QEALAPFRGK ELQPGSHIQS DKEIDAFVRA KADSAYHPSC TCKMGQPSDP TAVVDPQTRV LGVENLRVVD ASIMPSMVSG NLNAPTIMIA EKAADIIKGQ PALWDKDVPV YKPRTLATQR; Positive Samples: 293T, HepG2, Mouse thymus, Mouse kidney, Mouse liver, Mouse pancreas, Mouse testis, Rat kidney; Cellular Location: Mitochondrion inner membrane.
The protein encoded by this gene is a choline dehydrogenase that localizes to the mitochondrion. Variations in this gene can affect susceptibility to choline deficiency. A few transcript variants have been found for this gene, but the full-length nature of only one has been characterized to date.
Specifications
CHDH | |
Polyclonal | |
Unconjugated | |
CHDH | |
AU041472; CDH; CHD; Chdh; choline dehydrogenase; choline dehydrogenase, mitochondrial; D630034H06Rik | |
Rabbit | |
Affinity Chromatography | |
RUO | |
218865, 290551, 55349 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q6UPE0, Q8BJ64, Q8NE62 | |
CHDH | |
Recombinant fusion protein containing a sequence corresponding to amino acids 245-594 of human CHDH (NP_0608672). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.