Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CHDI 113
GREENER_CHOICESDP

Catalog No. 80635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-635 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 80-635 Supplier Tocris Bioscience™ Supplier No. 8063/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent HTT pre-RNA splicing modulator

CHDI 113 is a potent huntingtin (HTT) pre-mRNA splicing modulator (EC50 = 66 nM). It lowers HTT protein in both human Huntington's disease (HD) stem cells and in the brain of BACHD (bacterial artificial chromosome-mediated transgenic) mouse model. CHDI 113 is brain penetrant and orally bioavailable.

Specifications

Chemical Name or Material 5-[(3R)-3-(Cyclopropylamino)-1-pyrrolidinyl]-N-(8-fluoro-2-methylimidazo[1,2-a]pyridin-6-yl)-2-pyrazinecarboxamide
CAS 2743336-70-9
Quantity 5 mg
Target Huntingtin
Molecular Formula C20H22FN7O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.