Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral wnvNS1 Protein Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB290722SP
This item is not returnable.
View return policy
Description
wnvNS1 Protein Monoclonal specifically detects wnvNS1 Protein in Viral samples. It is validated for ELISA.Specifications
ELISA | |
991551 | |
ELISA | |
NP_776015 | |
HEK293 human embryonic kidney cell line transfected with recombinant Viral wnvNS1 Protein, Asp1-Ala352, Accession # NP_776015 | |
25 μg | |
Detects recombinant Viral wnvNS1 Protein in direct ELISAs. | |
Virus | |
Supernatant |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction