Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHEK1, Rabbit anti-Human, APC, Clone: F10, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB18854
Specifications
CHEK1 | |
Monoclonal | |
APC | |
In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
CHK1 | |
Rabbit | |
10 Reactions | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
F10 | |
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user. | |
CHK1 checkpoint homolog (S. pombe) | |
CHEK1 | |
A synthetic phosphopeptide corresponding to residues surrounding S317 of human CHEK1. | |
RUO | |
1111 | |
Store at 4°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction