Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEK1, Rabbit anti-Human, APC, Clone: R3F9, Abnova™

Catalog No. 89335700 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-700 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-700 Supplier Abnova Corporation Supplier No. MAB18866
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen CHEK1
Applications Flow Cytometry
Classification Monoclonal
Clone R3F9
Conjugate APC
Dilution Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Gene CHK1 checkpoint homolog (S. pombe)
Gene Alias CHK1
Gene Symbols CHEK1
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding S345 of human CHEK1.
Quantity 10 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1111
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.