Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEK2 Mouse anti-Canine, Human, Rat, Clone: OTI2F4, liquid, TrueMAB™
SDP

Catalog No. 501671539
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1539 Supplier OriGene Supplier No. TA500405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Checkpoint kinase 2 (Chk2) is a serine/threonine-protein kinase which functions as a cell cycle checkpoint regulator. ATM and ATR kinases are activated in response to ionizing and ultraviolet radition, and phosphorylate Thr68 and other sites in the amino-terminal region of Chk2 dimerisation and activation. Activated Chk2 phosphorylates and inhibits Cdc25C phosphates, preventing the entry into mitosis, and has been shown to stabilize the tumor suppressor protein p53, leading to cell cycle arrest in G1. In addition, Chk2 can phosphorylate BRCA1, allowing BRCA1 to restore survival after DNA damage. Chk2 is a putative tumor suppressor protein, with mutations in to the Chk2 gene linked to Li-Fraumeni syndrome, a familiar cancer usually associated with inherited mutations in TP53. Also, mutations in this gene are thought to confer a predisposition to sarcomas, breast cancer, and brain tumors.

Specifications

Antigen CHEK2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI2F4
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CHEK2
Gene Accession No. O96017
Gene Alias CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1
Gene Symbols CHEK2
Host Species Mouse
Immunogen Full length human recombit protein of human CHEK2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11200, 114212, 486338
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.