Learn More
Sigma Aldrich Chelex™ 100 Sodium Form
Catalog No. 95577100GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
50 mg
Catalog No. 95-577-100GF
Supplier: Sigma Aldrich 95577100GF
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Chelex 100 is a chelating resin. It can be used for the separation of uranium from fresh waters. Chelex 100 sodium form is the sodium form of purified chelex-100.Chelex™ 100 sodium form may be used in the removal of trace metals from artificial and natural seawater without changing the major cation-anion composition or contributing organic toxicants or chelators. It may be used in 2-deoxy-D-ribose degradation assay.Chelex is a registered trademark of Bio-Rad Laboratories, Inc.
Specifications
Quantity 50 mg
Synonym L-Proline 2-chlorotrityl ester polymer-bound
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.