Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chelidonic acid monohydrate, 96%

Catalog No. AC154600050
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic bottle

CAS: 6003-94-7 | C7H4O6·H2O | 202.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6003-94-7
Molecular Formula C7H4O6·H2O
Molecular Weight (g/mol) 202.12
MDL Number MFCD00149433
InChI Key KJZJAKSEIQOKAK-UHFFFAOYSA-N
Synonym chelidonic acid monohydrate, compound xi* hydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid monohydrate, chelidonic acidmonohydrate, chelidonic acid 1-hydrate, c7h4o6.h2o, 2,6-dicarboxy-4-oxo-4h-pyran monohydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid hydrate, 4h-pyran-2,6-dicarboxylicacid, 4-oxo-, hydrate 1:1
PubChem CID 51346158
IUPAC Name 4-oxopyran-2,6-dicarboxylic acid;hydrate
SMILES C1=C(OC(=CC1=O)C(=O)O)C(=O)O.O

Specifications

Melting Point 265°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 18, 490
Merck Index 14, 2052
Formula Weight 202.12
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Chelidonic acid monohydrate, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.