Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chemerin Antibody (13G2), PerCP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP221989PCP
Description
Chemerin Monoclonal antibody specifically detects Chemerin in Mouse samples. It is validated for ELISASpecifications
Chemerin | |
Monoclonal | |
PerCP | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
13G2 | |
ELISA | |
chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433 | |
Mouse recombinant Chemerin | |
0.1 mL | |
Cardiovascular Biology, metabolism | |
5919 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction