Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chemerin Polyclonal Antibody
Chemerin Polyclonal Antibody
Supplier: Thermo Scientific PA575669
Description
Chemerin Polyclonal Antibody for Western Blot

Specifications
Chemerin | |
Polyclonal | |
Unconjugated | |
Rarres2 | |
0610007L05Rik; AI303516; Chemerin; HP10433; Rarres2; RAR-responsive protein TIG2; retinoic acid receptor responder (tazarotene induced) 2; retinoic acid receptor responder 2; retinoic acid receptor responder protein 2; Tazarotene-induced gene 2 protein; TIG2; TIG-2; TIG2HP10433 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
297073, 5919, 71660 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99969, Q9DD06 | |
Rarres2 | |
Synthetic peptide corresponding to residues in Human Chemerin. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction