Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHEMetrics CHEMets Dissolved Oxygen Test Kit Refill

Click to view available options
Range:
0 to 1 ppm
0 to 40 ppb and 0 to 100 ppb
1 to 12 ppm
Test Method:
Indigo Carmine
Rhodazine D
For Use With (Equipment):
C-7501 comparators
C-7512 comparators
C-7540 comparators
Description
- Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
- Comparators have two-year shelf life
Specifications
Specifications
| Analytes | Dissolved Oxygen (DO) |
| For Use With (Equipment) | C-7501 comparators |
| Includes | 30 self-filling CHEMets ampoules to refill kit K-7501 |
| No. of Tests | 30 |
| Range | 0 to 1 ppm |
| Test Method | Rhodazine D |
| Test Time | 2 min. |
| Type | CHEMets Dissolved Oxygen Refill |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction