CHEMetrics™ Chemets™ Water Testing Kits
Provide results in two minutes or less
Manufacturer: CHEMetrics™ K7512
 View more versions of this product
Catalog No. 13-299-200
  Check Availability
Add to Cart
    Specifications
    Includes Plastic case, 30 ampoules of reagent, refill, comparator, 25mL sample cup, instructions
    No. of Tests 30.0
    Test Time 2 min.
    Method Indigo Carmine
    Analytes Dissolved Oxygen
    Product Type Chemets™ Water Testing Kit
    For Use With (Application) Water testing kits for chlorine (DPD), copper, dissolved oxygen, hydrogen peroxide, phenols, phosphate, silica, sulfide and more
    Range 1 to 12ppm
  • Description & Specifications
Related Products
Product Content Feedback