Learn More
CHEMetrics COD Vials Kits

Description
Includes
Instructions
Requires
Requires the use of a Digestor Block and the V-2000 Photometer, a COD Photometer, or a spectrophotometer capable of accepting a 16mm round cell. Instruments sold separately.
- Contains everything needed (except distilled water) to perform tests
- Includes vials and instruction book
- Comes in cardboard box
- 2-hour digestion time
- Dichromate reactor digestion
A fresh reagent ampule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.
Certifications
USEPA-accepted
Specifications
Specifications
| Analytes | Chemical Oxygen Demand (COD) |
| Certifications/Compliance | USEPA accepted |
| For Use With (Application) | Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater |
| For Use With (Equipment) | Digestor block as well as a photometer or spectrophotometer |
| Includes | 150 vials and instruction book |
| No. of Tests | 149 |
| Range | 0 to 150 ppm |
| Test Method | Dichromate reactor digestion |
| Test Time | 2 hr. digestion |
| Type | COD Vials Kit |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.