Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHEMetrics COD Vials Kits

Click to view available options
Range:
0 to 15,000 ppm
0 to 15,000 ppm (HR+)
0 to 150 ppm
0 to 1500 ppm
Certifications/Compliance:
Not USEPA approved
USEPA accepted
Includes:
150 vials and instruction book
25 vials and instruction book
98 vials and instruction book
No. of Tests:
24
25
149
Description
Includes
Instructions
Requires
Requires the use of a Digestor Block and the V-2000 Photometer, a COD Photometer, or a spectrophotometer capable of accepting a 16mm round cell. Instruments sold separately.
- Contains everything needed (except distilled water) to perform tests
- Includes vials and instruction book
- Comes in cardboard box
- 2-hour digestion time
- Dichromate reactor digestion
A fresh reagent ampule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.
Certifications
Not USEPA Approved
Specifications
Specifications
| Analytes | Chemical Oxygen Demand (COD) |
| Certifications/Compliance | Not USEPA approved |
| EPA Approved Methods | Not USEPA approved |
| For Use With (Application) | Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater |
| For Use With (Equipment) | Digestor block as well as a photometer or spectrophotometer |
| Includes | 98 vials and instruction book |
| Range | 0 to 15,000 ppm |
| Test Method | Dichromate reactor digestion |
| Test Time | 2 hr. digestion |
| Type | COD Vials Kit |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction