Learn More
CHEMetrics CHEMets Chlorine Free & Total Water Testing Kit
SDP
Catalog No. p-7238968
Click to view available options
Range:
0 to 1 ppm and 1 to 5 ppm
0.1 to 2 ppm
Test Method:
DDPD
DPD
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
REMOVE
Provides results in two minutes or less
Provide results in two minutes or less
  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life
Specifications
Analytes Free and Total Chlorine
Includes Plastic case, 30 ampoules of reagent, comparator, snap cup and instructions
No. of Tests 30
Range 0 to 1 ppm and 1 to 5 ppm
Test Method DDPD
Type CheMets Water Testing Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.