Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Chemical, Biohazard & Hazardous Material Label - Self-Sticking Polyester
Shop All Brady Corporation Products
Click to view available options
Display Text:
ACETYLENE
CHAIN CYLINDERS
CORROSIVE MATERIALS
DANGER DIESEL FUEL
DIESEL FUEL
FLAMMABLE GAS
FLAMMABLE MATERIALS
GASOLINE
LEAD WORK AREA POISON NO SMOKING OR EATING
OXYGEN
Description
- Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
- Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
- Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
- Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
Specifications
Autoclavable | Not Autoclavable |
Certifications/Compliance | Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200 |
Description | Label |
Adhesive Type | Permanent Acrylic |
Color | Red on White |
Finish | Gloss |
Format | Self-adhesive |
Material | Polyester |
Printable Sides | Single-side Printable |
Product Type | Label |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction