Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Chemical, Biohazard & Hazardous Material Labels - Self-Sticking Polyester - 3.5 in H x 5 in W

Catalog No. p-7111387 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
ALWAYS
BIOLOGICAL
CAUSTIC
CAUTION.THIS EQUIP
CHEMICAL STORAGE
CORROSIVE
CORROSIVE MATE
DANGER Acid
DANGER CONFINED
DANGER FOLLOW
DANGER HIGH
DANGER NEVER REACH
DANGER WATCH YOUR HANDS AND FINGERS
DANGER.DEFECTIVE
DANGER.DO NOT
DANGER.TURN OFF
DANGER.WATCH YOUR
DANGEROUS
DO NOT OPERATE WITHOUT GUARDS IN PLACE
EXPLOSIVES
FLAMMABLE
INHALATION
NOTICE AUTHORIZED
PELIGRO CALIENTE
PELIGRO INFLAMMABLE
WARNING HOT SURFACE DO NOT TOUCH
WARNING.FLAMMABLE
WARNING.THIS MACHINE
29 product options available for selection
Product selection table with 29 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
19-107-539 CORROSIVE
19-107-540 DANGEROUS
19-107-541 DANGER.DEFECTIVE
19-107-544 PELIGRO INFLAMMABLE
19-107-542 EXPLOSIVES
19-107-545 INHALATION
19-107-546 DANGER.TURN OFF
19-107-547 DANGER.WATCH YOUR
19-107-548 BIOLOGICAL
19-107-549 FLAMMABLE
19-107-551 ALWAYS
19-107-553 CORROSIVE MATE
19-107-552 CHEMICAL STORAGE
19-107-555 CAUTION.THIS EQUIP
19-107-556 DANGER HIGH
19-107-557 DO NOT OPERATE WITHOUT GUARDS IN PLACE
19-107-559 NOTICE AUTHORIZED
19-107-558 DANGER.DO NOT
19-107-560 DANGER NEVER REACH
19-107-561 DANGER WATCH YOUR HANDS AND FINGERS
19-107-563 DANGER FOLLOW
19-107-562 DANGER CONFINED
19-107-564 WARNING.FLAMMABLE
19-107-565 WARNING.THIS MACHINE
19-107-570 PELIGRO INFLAMMABLE
19-107-568 CAUSTIC
19-107-567 DANGER Acid
19-107-571 PELIGRO CALIENTE
19-107-550 WARNING HOT SURFACE DO NOT TOUCH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
29 options
Catalog No. 19-107-539 Supplier Brady™ Supplier No. 86123

Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.

  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200
Description Label
Adhesive Type Permanent Acrylic, Self-adhesive
Finish Gloss
Format Label
Material Polyester
Printable Sides Single-side Printable
Product Type Label
Display Text CORROSIVE
Languages English
Quantity 5/Pk.
Shape Rectangle
Size (English) 5 x 3.5 in. (W x H)
Dimensions (LxW) 3.5 x 5 in.
Width (English) 5 in.
Width (Metric) 92.71 mm
Height (English) 3.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.