Learn More
Description
- Withstand most chemicals, solvents and vapors
- Resist fading and scratching
- For medium-sized containers
- Measure 3 × 5 in.
Specifications
Specifications
| Color | White, Red |
| Format | Label |
| Material | Vinyl |
| Product Type | Kerosene Container Label |
| For Use With (Application) | Hazard identification, suitable for both indoor and outdoor applications |
| Quantity | 10/Pk. |
| Thickness (English) | 0.0045 mil |
| Width (English) | 5 in. |
| Height (English) | 3 in. |
| Height (Metric) | 7.62 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.