Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Chemical Hazard Identification Labels

Catalog No. 1798160C Shop All National Marker Company Products
Change view
Click to view available options
Product Type:
Acetone Container Label
Bleach solid Container Label
Diesel fuel Container Label
Ethyl alcohol Container Label
Gasoline Container Label
Hydrochloric acid Container Label
Isopropyl alcohol Container Label
Kerosene Container Label
Methyl ethyl ketone Container Label
Sulfuric acid Container Label
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-981-60C Methyl ethyl ketone Container Label
17-981-59T Hydrochloric acid Container Label
17-981-59E Ethyl alcohol Container Label
17-981-59A Acetone Container Label
19-035-036 Diesel fuel Container Label
17-981-59W Isopropyl alcohol Container Label
17-981-59R Gasoline Container Label
17-981-59Z Kerosene Container Label
19-035-035 Bleach solid Container Label
17-981-60N Sulfuric acid Container Label
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17-981-60C Supplier National Marker™ Supplier No. HC 271

High-visibility red-and-white pressure-sensitive vinyl labels ideal for alerting employees to potential chemical hazards in the workplace

  • Withstand most chemicals, solvents and vapors
  • Resist fading and scratching
  • For medium-sized containers
  • Measure 3 × 5 in.

Specifications

Color White, Red
Format Label
Material Vinyl
Product Type Methyl ethyl ketone Container Label
For Use With (Application) Hazard identification, suitable for both indoor and outdoor applications
Quantity 10/Pk.
Thickness (English) 0.0045 mil
Width (English) 5 in.
Height (English) 3 in.
Height (Metric) 7.62 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.