Learn More
Innovating Science™ Bacteria Stain Chemical Kit
GREENER_CHOICESCI_ED
Catalog No. s98664 Shop All Aldon Products
Click to view available options
Quantity:
1 Kit
WARNING: This product can expose you to chemicals including Crystal Violet and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
WARNING: This product can expose you to chemicals including Crystal Violet and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
Identify and differentiate various types of bacteria using this bacteria stain kit.
Kit Includes:
  • 30 mL Methylene Blue Loefflers Solution
  • 30 mL Methylene Blue Saturated, 1% in IPA/ Water
  • 30 mL Carbol Fuchsin Solution
  • 30 mL Carbol Rose Bengal Solution
  • 30 mL Crystal Violet, 1% Alcohol
Specifications
Type Bacteria Stain Kit
Quantity 1 Kit
Certifications/Compliance 49 CFR 173.4
Includes 30mL methylene blue Loefflers solution, 30mL methylene blue Saturated, 30mL 1% in IPA or water, 30mL Carbol Fuchsin solution, 30mL Carbol Rose Bengal solution, 30mL Crystal Violet, 30mL 1% alcohol
Volume (Metric) 5 x 30 mL
Sterility Non-sterile
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.