Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Adult Rat Hippocampal Neural Stem Cells

Catalog No. scr022 Shop All MilliporeSigma Products
Click to view available options
:
Adult rat hippocampal neural stem cells

Catalog No. SCR022

Supplier: MilliporeSigma™ SCR022

Only null left
Add to Cart
Add to Cart

For stem cell culture applications

  • Self-renewing cells with capacity to differentiate into neurons, astrocytes and oligodendrocytes
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen
Cell Type Neural Stem Cells
For Use With (Application) Stem cell culture
Quantity 1 vial

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.