Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Dextran Sulfate 50% Solution

Catalog No. p-7078318 Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 mL
200 mL

Used as additive in probe hybridization solutions.

  • Presence accelerates rate of hybridization by 10 fold
  • Favors formation of probe networks
  • Greater sensitivity during membrane bound nucleic acid analysis
  • 50% (w/v) solution
TRUSTED_SUSTAINABILITY

Specifications

Includes Mitochondria Extraction Buffer, Cytosol Extraction Buffer, Protease Inhibitor Cocktail, Reducing Reagent, and user protocol.
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.