Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Fibronectin α Chymotryptic Fragment (Heparin-binding region), Human Purified

Catalog No. p-5393999 Shop All MilliporeSigma Products
Click to view available options
Product Type:
Fibronectin 120 kd α Chymotryptic Fragment
Fibronectin 40 kd α Chymotryptic Fragment

Catalog No. F1904

Supplier: MilliporeSigma™ F1904

Only null left
Add to Cart
Add to Cart

For Cell Culture applications

  • Heparin-binding fragment promotes cytoskeleton organization in integrin signaling
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) Cell Culture
pH Range 7.4
Quantity 500 μg
Source Human plasma cryoprecipitate (plasma fibronectin), assayed negative for hepatitis B virus, hepatitis C Virus (HCV), HIV-1, HIV-2, HTLV-1, HTLV-2 and Treponema pallidum.
Product Type Fibronectin 120 kd α Chymotryptic Fragment
Content And Storage Maintain material at -20°C for 3 months or -70°C for six months.
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.