Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Chemicon™ Fibronectin α Chymotryptic Fragment (Heparin-binding region), Human Purified
Shop All MilliporeSigma Products
Click to view available options
Product Type:
Fibronectin 120 kd α Chymotryptic Fragment
Fibronectin 40 kd α Chymotryptic Fragment
Description
- Heparin-binding fragment promotes cytoskeleton organization in integrin signaling
Specifications
Specifications
| Concentration | 1 mg/mL |
| For Use With (Application) | Cell Culture |
| pH Range | 7.4 |
| Quantity | 500 μg |
| Source | Human plasma cryoprecipitate (plasma fibronectin), assayed negative for hepatitis B virus, hepatitis C Virus (HCV), HIV-1, HIV-2, HTLV-1, HTLV-2 and Treponema pallidum. |
| Product Type | Fibronectin 120 kd α Chymotryptic Fragment |
| Content And Storage | Maintain material at -20°C for 3 months or -70°C for six months. |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction