Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Human Collagen Type III

Catalog No. CC054
Click to view available options
Quantity:
100 μg

Adds structure and strength to connective tissues

  • COL3A1 is gene responsible for producing the pro-alpha1(III) chain of type III procollagen
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) Cell Culture
pH Range 2.5
Quantity 100 μg
Product Type Collagen, Human Type III
Content And Storage Maintain at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.
Form Liquid
Purity or Quality Grade 90% pure by SDS-PAGE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.