Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Chemicon™ Human Plasma Fibronectin Purified Protein
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
100 mg
Description
- Used as attachment factor
Specifications
Specifications
| Concentration | 1 mg/mL |
| For Use With (Application) | Cell Culture |
| Formulation | Lyophilized from 150 mM NaCl, 10 mM sodium phosphate, pH 7.5, sterile filtered, containing no preservatives. |
| pH Range | 7.5 |
| Quantity | 100 mg |
| Source | Human Donor Plasma |
| Product Type | Human Plasma Fibronectin Purified Protein |
| Content And Storage | Maintain at 2° to 8°C for up to 6 months from date of receipt. Do not freeze. |
| Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction