Learn More
MilliporeSigma&trade Chemicon™ Human Plasma Fibronectin Purified Protein Available on GSA/VA Contract for Federal Government customers only.
Suitable for cell propagation in vitro
Manufacturer:  MilliporeSigma&trade FC010
 View more versions of this product
Catalog No. FC010
Add to cart
Description
Description
  • Used as attachment factor
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg/mL
Liquid
Maintain at 2° to 8°C for up to 6 months from date of receipt. Do not freeze.
Human donor plasma
7.5
Cell culture
Liquid in 150 mM NaCl, 10 mM sodium phosphate, pH 7.5, sterile filtered, containing no preservatives.
1mg
Human plasma fibronectin purified protein