Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Chemicon™ Recombinant Human Leukemia Inhibitory Factor Protein
Click to view available options
Quantity:
10 μg
5 μg
Description
Leukaemia inhibitory factor is a cytokine that induces macrophage differentiation. Neurotransmitters and neuropeptides, well known for their role in the communication between neurons, are also capable of activating monocytes and macrophages and inducing chemotaxis in immune cells. LIF signals through different receptors and transcription factors. LIF in conjunction with BMP2 acts in synergy on primary fetal neural progenitor cells to induce astrocytes.

Specifications
Specifications
Content And Storage | 2- 8°C for 6 - 12 months |
For Use With (Application) | Cell culture |
Quantity | 5 μg |
Source | E. coli |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction