Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Chemiluminescence in Blood Stain Detection Kit
Shop All Aldon Products

Click to view available options
Class Size:
15 Lab Groups
Description
Includes
Instruction manual with suggested activities, two bottles of Luminol, rehydrating solution, Luminol powder, simulated blood hemoglobin
- Crime scene investigators are called to the scene of a possible violent crime.
- Examine the scene for evidence including fingerprints, hair and fibers.
- After collecting evidence, they spray the area with a liquid and turn out the lights. A strange, faint glow appears in certain areas of the scene.
- Special Luminol formulation does not require a separate hydrogen peroxide catalyst.
- Simply rehydrate and use with simulated blood hemoglobin.
- Enough material for several demonstrations.
Specifications
Specifications
Color | White |
Grade | 7 to 14 |
Age | 13+ years |
For Use With (Application) | To learn how luminol is used in crime scenes |
Includes | Instruction manual with suggested activities, two bottles of luminol, rehydrating solution, luminol powder, simulated blood hemoglobin |
Quantity | 1 |
Dimensions (L x W x H) | 9.75 x 6.75 x 3 in. |
Class Size | 15 Lab Groups |
Product Type | Blood Stain Detection Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction