Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Chemiluminescence Chemical Reaction Kit
GREENER_CHOICESCI_ED

Catalog No. s73016 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S73016 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S73016 Supplier Innovating Science™ Supplier No. IS7013
Only null left
Add to Cart
Add to Cart

Includes

Materials for five demonstrations: 100 mL Luminol Solution; 50 mL Hydrogen Peroxide, 6% Solution; and instructions

Explain and view chemiluminescence.

  • Observe how luminol is converted to an excited state in the presence of an oxidizer such as hydrogen peroxide
  • As the excited-state molecule decays or returns to the ground state, energy is produced as light

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) For chemiluminescence chemical reaction
Includes 1 x 100 mL luminol solution, 1 x 50 mL hydrogen peroxide, 6% solution
Quantity 1
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 1 Class
Product Type Chemiluminescence Demonstration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.