Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Chemiluminescent Nucleic Acid Detection Module Kit PROMO

Catalog No. PI89880PM
Click to view available options
Quantity:
10 Blots

Detect biotin-labeled nucleic acid probes in various membrane blotting applications with this complete chemiluminescent detection system.

The Thermo Scientific Chemiluminescent Nucleic Acid Detection Module is a complete system for detection of biotin-labeled nucleic acids for various blotting applications including Northern/Southern blots, ribonuclease protection assays (RPAs) and electrophoretic mobility shift assays (EMSAs). This module provides the essential detection components used in the Thermo Scientific North2South and LightShift EMSA Kits. The system includes an enhanced luminol substrate for horseradish peroxidase (HRP) with optimized blocking and wash buffers that together produce sensitivity equivalent to radioactive (32P) systems. Obtain publication-quality blots having excellent signal and low background.

Highlights:

  • Fast and sensitive – achieve the same speed and detection levels you've come to expect with the North2South and LightShift Kits

  • Versatile – kit protocol (included) is applicable to any biotin-based nucleic acid detection system: Northerns, Southerns, gel shifts, RPAs and colony lifts

  • Flexible – kit contains only the critical detection components for those who already have biotin-labeled probes and an optimized hybridization method developed

Includes:

Stabilized Streptavidin-HRP Conjugate 1.5 mL, Luminol/Enhancer Solution 80 mL, Stable Peroxide Solution 80 mL, Blocking Buffer 500 mL, 4X Wash Buffer 500 mL, Substrate Equilibration Buffer 500 mL

Specifications

Content And Storage Sufficient For: 800 cm2 of membrane (10 mini-gel blots), Stabilized Streptavidin-HRP, 1.5 mL, Luminol/Enhancer Solution, 80 mL, Stable Peroxide Solution, 80 mL, Blocking Buffer (Part No. 89880A), 500 mL, Wash Buffer (4X), 500 mL, Substrate Equilibration Buffer, 500 mL Store at 4°C.
Detection Method Chemiluminescence
Format Kit
Product Line Pierce™
Product Type Nucleic Acid Detection Module Kit
Quantity 10 Blots

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.