Learn More
Thermo Scientific™ Cheminert Model C2 Microbore Injector
Can be used as an injector or switching valve
Catalog Number | Mfr. No. | Volume (Metric) Sample | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) Sample | Price | Quantity & Availability | |||||
03-052-416
|
Thermo Scientific™
CZSL100PK |
100 μL | N/A | N/A | N/A | ||||
Description
- 1/16 in. fittings
- 0.010 in. ports
- Available in 6-port or 10-port configurations
- Available with manual or microelectric actuator