Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Chemistry Demonstration Kits: Reduction-Oxidation Rainbow
GREENER_CHOICESCI_ED

Catalog No. S68307 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S68307 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.
Catalog No. S68307 Supplier Innovating Science™ Supplier No. IS7027
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.

Rainbow reaction colorfully demonstrates an oscillation reaction.

In this demonstration, hydrogen peroxide is added to a colorless solution of potassium iodide and a red color is produced. But when hydrogen peroxide is added to a purple solution of potassium permanganate, the solution becomes colorless! What causes these dramatically different color changes? The explanation lies in the fact that hydrogen peroxide can behave as an oxidizing agent or a reducing agent, depending on the circumstances. Hydrogen peroxide oxidizes iodide ion to iodine. However, in the presence of a strong oxidizer such as potassium permanganate, hydrogen peroxide becomes a weak reducing agent, reducing the permanganate ion to the manganese ion plus oxygen.

Specifications

Certifications/Compliance DOT Info: Small quantity exemption 173.4. This package conforms TO 49 CFR 173.4 for domestic highway or rail transport only.
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Redox Reactions
Includes 1 x 100 mL hydrogen peroxide, 3% solution, 1 x 200 mL potassium iodide, 0.1 m solution, 1 x 200 mL potassium, permanganate, 0.02 m solution, 1 x 10 mL sulfuric acid, concentrated un1830
Quantity 1
Dimensions (L x W x H) 6 x 4.25 x 6.875 in.
Class Size 2 Lab Groups
Product Type Reduction-Oxidation Rainbow Demonstration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.