Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chemistry Problems

Click to view available options
Grade:
9 to 12
Description
- Work on problem-solving and proficiency in analytical and math skills
- Reproducible information on atomic structure, chemical reactions, stoichiometry and acid-base chemistry
- Complete answer key and references to applicable websites included
- David E. Newton, 2002, fifth edition, 220 pages
- Grades 9 to 12
Specifications
Specifications
Grade | 9 to 12 |
Format | Book |
Product Type | Chemistry Problems |
Includes | Complete answer key and reference to applicable web sites |
For Use With (Application) | Chemistry problems and experiments |
Subject | Chemistry |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction