Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemR23/CMKLR1 Antibody (84939), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB312849 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312849 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312849 Supplier Novus Biologicals Supplier No. FAB362PECY7
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen ChemR23/CMKLR1
Applications Flow Cytometry
Classification Monoclonal
Clone 84939
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human ChemR23, Met1-Leu371, Accession # NP_004063
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1240
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.