Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChemR23/CMKLR1 Antibody (BZ194), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP311985AF532
Description
ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
ChemR23/CMKLR1 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rat | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
IgG2a κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
BZ194 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez | |
This antibody was raised by immunising Wistar Furth rats with the mChemR23/CMKLR1 peptide/KLH conjugate. The immunizing amino-terminal mChemR23/CMKLR1 peptide with the sequence NH2-DSGIYDDEYSDGFGYFVDLEEASPWC-COOH, corresponding to residues residues 8-32 of mChemR23/CMKLR1, with a nonnative carboxyl-terminal cysteine to facilitate conjugation to keyhole limpet hemocyanin (KLH). | |
0.1 mL | |
GPCR, Immunology | |
1240 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction