Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChemR23/CMKLR1 Antibody (BZ194) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280667
Description
ChemR23/CMKLR1 Monoclonal specifically detects ChemR23/CMKLR1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ChemR23/CMKLR1 | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry : 1:100 - 1:200, Immunohistochemistry-Paraffin | |
CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
BZ194 | |
Unconjugated | |
PBS | |
CMKLR1 | |
The immunogen for this antibody was CMKLR1. | |
0.1 mg | |
GPCR, Immunology | |
1240 | |
Store at 4°C. Do not freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction