Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChemR23/CMKLR1 Antibody (BZ194), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143233MFV610
Description
ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Mouse samples. It is validated for Flow CytometrySpecifications
ChemR23/CMKLR1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
BZ194 | |
Flow Cytometry | |
CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez | |
Partial synthetic peptide made to an N-terminal portion of the mouse ChemR23/CMKLR1 protein (amino acids 8-32) [UniProt P97468] | |
0.1 mL | |
GPCR, Immunology | |
1240 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction