Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ChemR23 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB362N100UG
This item is not returnable.
View return policy
Description
ChemR23/CMKLR1 Monoclonal specifically detects ChemR23/CMKLR1 in Human samples. It is validated for Flow Cytometry.Specifications
| ChemR23/CMKLR1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_004063 | |
| CMKLR1 | |
| NS0 mouse myeloma cell line transfected with human ChemR23 Met1-Leu371 Accession # NP_004063 | |
| RUO | |
| 1240 | |
| Human | |
| IgG3 |
| Flow Cytometry | |
| 84939 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, cmklr1, Dez, Gpcr27, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ChemR23. Stains human ChemR23-transfected cells but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction