Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Serged Seams, Attached Hood, Elastic Wrists and Ankles
Catalog No. 17184131 Shop All International Enviroguard Systems Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
These durable and disposable coveralls with serged seams and elastic back protect against chemical splashes and toxic particles.
  • Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
  • A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
  • A high-contrast red stripe lets you quickly distinguish this taped-seam garment from coveralls made with serged seams
  • For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
  • Elastic wrists and ankles provide a secure closure that helps keep pants and sleeves in place to prevent skin exposure
  • Roomy, seamless shoulder area resists tears to provide more protection in high-movement areas
  • A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
  • Elastic back stretches with user for more comfort and better tear resistance
  • High moisture vapor transfer rate (MVTR) allows heat and moisture vapor to escape, so heat isn't trapped near the body
  • Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
Product Type Chemical Protective Clothing
Size 2X-Large
Gender Unisex
Material Laminated Non-woven Fabric
Color Yellow
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Finger Loops
Neck Style Collared
Hood Type Attached, Elastic Face
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Resistance Acid, Caustics, Chemicals, Isopropyl Alcohol, Particle
Latex Free Yes
No. of Pockets None
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.