Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Taped Seams, Elastic Wrist and Ankle
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
- A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
- A high-contrast red stripe lets you quickly distinguish this taped-seam garment from coveralls made with serged seams
- For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
- Roomy, seamless shoulder area resists tears and provides more protection in key exposure area
- A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- Storm flap over zipper with pressure-sensitive tape adds an extra layer of protection
- High moisture vapor transfer rate improves wearer comfort
- Elastic wrists offer a secure fit to keep sleeves in place to prevent skin exposure
- Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
Specifications
Product Type | Chemical Protective Clothing |
Size | Medium |
Gender | Unisex |
Material | Laminated Non-woven Fabric |
Color | Yellow |
Disposable | Single-use |
Waist Style | Elastic at Back |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Collared |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction